Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XKR6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555198
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555198 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555198 Supplier Invitrogen™ Supplier No. PA555198
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YGVLHPTGPR AKILASSCCA ELLWGIPLPP DVEPMAPEIP GYRGTQVTPT RAVTEQQEDL TADTCLPVFQ VRPMGPPTPL GRPYLPEGPL IKIDMPR Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

XK is a 444 amino acid protein that spans the membrane 10 times and carries the ubiquitous antigen, Kx, which determines blood type. The XK (X-linked Kx blood group)-related gene family are homologs of XK. XKR6 (XK-related protein 6) is a 641 amino acid multi-pass membrane protein that likely is a component of the XK/Kell complex of the Kell blood group system. The gene encoding XKR6 maps to human chromosome 8, which is made up of nearly 146 million bases and encodes about 800 genes. There are two isoforms of XKR6 that are produced as a result of alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XKR6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene XKR6
Gene Accession No. Q5GH73
Gene Alias BC024502; C8orf21; C8orf5; C8orf7; LOW QUALITY PROTEIN: XK-related protein 6; RGD1310719; Transmembrane protein C8orf5; X Kell blood group precursor related family member 6 homolog; X Kell blood group precursor-related family, member 6; XK related 6; XK, Kell blood group complex subunit-related family, member 6; XKR6; XK-related protein 6; X-linked Kx blood group related 6; Xrg6
Gene Symbols XKR6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human XKR6. Recombinant protein control fragment (Product #RP-93900).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286046
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.