Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XKR6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570774
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570774 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570774 Supplier Invitrogen™ Supplier No. PA570774
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 86%; Rat: 93%; Zebrafish: 79%.

XK is a 444 amino acid protein that spans the membrane 10 times and carries the ubiquitous antigen, Kx, which determines blood type. The XK (X-linked Kx blood group)-related gene family are homologs of XK. XKR6 (XK-related protein 6) is a 641 amino acid multi-pass membrane protein that likely is a component of the XK/Kell complex of the Kell blood group system. The gene encoding XKR6 maps to human chromosome 8, which is made up of nearly 146 million bases and encodes about 800 genes. There are two isoforms of XKR6 that are produced as a result of alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XKR6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene XKR6
Gene Accession No. Q5GH73
Gene Alias BC024502; C8orf21; C8orf5; C8orf7; LOW QUALITY PROTEIN: XK-related protein 6; RGD1310719; Transmembrane protein C8orf5; X Kell blood group precursor related family member 6 homolog; X Kell blood group precursor-related family, member 6; XK related 6; XK, Kell blood group complex subunit-related family, member 6; XKR6; XK-related protein 6; X-linked Kx blood group related 6; Xrg6
Gene Symbols XKR6
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human XKR6.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286046
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.