Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XKR7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA551271
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA551271 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA551271 Supplier Invitrogen™ Supplier No. PA551271
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total XKR7 protein.

XKR7 (XK-related protein 7) is a 579 amino acid multi-pass membrane protein that likely is a component of the XK/Kell complex of the Kell blood group system. The gene encoding XKR7 maps to human chromosome 20, which comprises approximately 2% of the human genome. Chromosome 20 contains nearly 63 million bases that encode over 600 genes, some of which are associated with Creutzfeldt-Jakob disease, amyotrophic lateral sclerosis, spinal muscular atrophy, ring chromosome 20 epilepsy syndrome and Alagille syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XKR7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene XKR7
Gene Accession No. Q5GH72
Gene Alias C20orf159; dJ310O13.4; X Kell blood group precursor-related family, member 7; XK related 7; XK, Kell blood group complex subunit-related family, member 7; XKR7; XK-related protein 7; X-linked Kx blood group related 7; XRG7
Gene Symbols XKR7
Host Species Rabbit
Immunogen Synthetic peptide of human XKR7.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 343702
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.