Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XKR8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598929
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598929 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598929 Supplier Invitrogen™ Supplier No. PA598929
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

XKR8 Polyclonal Antibody for Western Blot, ELISA

Promotes phosphatidylserine exposure on apoptotic cell surface, possibly by mediating phospholipid scrambling. Phosphatidylserine is a specific marker only present at the surface of apoptotic cells and acts as a specific signal for engulfment. Has no effect on calcium-induced exposure of phosphatidylserine. Activated upon caspase cleavage, suggesting that it does not act prior the onset of apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XKR8
Applications ELISA, Western Blot, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene XKR8
Gene Accession No. Q9H6D3
Gene Alias hXkr8; RP11-460I13.3; X Kell blood group precursor-related family, member 8; XK related 8; XK, Kell blood group complex subunit-related family, member 8; XKR8; XK-related protein 8; XK-related protein 8, processed form; X-linked Kx blood group related 8; XRG8
Gene Symbols XKR8
Host Species Rabbit
Immunogen Recombinant Human XK-related protein 8 protein (69-158AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55113
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.