Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XKR8 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281866
Description
XKR8 Polyclonal specifically detects XKR8 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| XKR8 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| XKR8 | |
| XKR8 antibody was raised against a 15 amino acid peptide near the carboxy terminus of human XKR8. The immunogen is located within the last 50 amino acids of XKR8. Amino Acid Squence: WKPDPDQVDGARSL | |
| Affinity Purified | |
| RUO | |
| 55113 | |
| Human | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| RP11-460I13.3, XKR8 XK, Kell blood group complex subunit-related family, member 8, XRG8 | |
| Rabbit | |
| 47 kDa | |
| 0.1 mg | |
| Primary | |
| XKR8 antibody is human reactive. At least two isoforms are known to exist; this antibody will detect both isoforms. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction