Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XKRX Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA551273

Catalog No. PA551273


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total XKRX protein.

This gene encodes a protein that is related to a component of the XK/Kell complex of the Kell blood group system. The encoded protein includes several transmembrane domains, is known to be exposed to the cell surface, and may function as a membrane transporter.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

XKRX
Polyclonal
Unconjugated
XKRX
membrane protein XPLAC; X Kell blood group precursor-related, X-linked; X Kell blood group-related, X-linked; XK related, X-linked; XK, Kell blood group complex subunit-related, X-linked; XKR2; XK-related protein 2; XKRX; X-linked Kx blood group related, X-linked; XPLAC; XRG2
Rabbit
Antigen affinity chromatography
RUO
402415
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1.6 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q6PP77
XKRX
Synthetic peptide of human XKRX.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.