Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XKRX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116244
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116244 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116244 Supplier Invitrogen™ Supplier No. PA5116244
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

XKRX Polyclonal Antibody for Western Blot

This gene encodes a protein that is related to a component of the XK/Kell complex of the Kell blood group system. The encoded protein includes several transmembrane domains, is known to be exposed to the cell surface, and may function as a membrane transporter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XKRX
Applications Western Blot
Classification Polyclonal
Concentration 0.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene XKRX
Gene Accession No. Q6PP77
Gene Alias membrane protein XPLAC; X Kell blood group precursor-related, X-linked; X Kell blood group-related, X-linked; XK related, X-linked; XK, Kell blood group complex subunit-related, X-linked; XKR2; XK-related protein 2; XKRX; X-linked Kx blood group related, X-linked; XPLAC; XRG2
Gene Symbols XKRX
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 317-345 amino acids from the C-terminal region of human XKRX.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 402415
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.