Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ XL 184
GREENER_CHOICESDP

Catalog No. 542250 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
54-225-0 50 mg
54-221-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 54-225-0 Supplier R&D Systems™ Supplier No. 5422/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent VEGFR inhibitor; also inhibits other RTKs

XL 184 is a potent VEGFR inhibitor (IC50 = 0.035 nM); also inhibits c-Met, KIT, RET, FLT4, AXL, FLT3, FLT1 and Tie2 (IC50 values are 1.3, 4.6, 5.2, 6, 7, 11.3, 12 and 14.3 nM, respectively). Induces intratumoral hypoxia and apoptosis. Reduces tumor invasion and metastasis in vivo. Antiangiogenic.

Specifications

Quantity 50 mg
Product Line Tocris
Product Type XL 184
Purity >98%
Molecular Weight (g/mol) 501.51
Inhibitors XL 184

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.