Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XLF Antibody (OTI1B5), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274880V
Description
XLF Monoclonal specifically detects XLF in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
XLF | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NHEJ1 | |
Full length recombinant protein of human XLF (NP_079058) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1B5 | |
Western Blot, Immunohistochemistry | |
Cernunnos, non-homologous end-joining factor 1, nonhomologous end-joining factor 1, Protein cernunnos, XLFFLJ12610, XRCC4-like factor | |
Mouse | |
Affinity Purified | |
Cancer, Chromatin Research, DNA Repair | |
79840.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction