Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ XLF Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA517198
Description
It is not recommended to aliquot this antibody.
Double-strand breaks in DNA result from genotoxic stresses and are among the most damaging of DNA lesions. This gene encodes a DNA repair factor essential for the nonhomologous end-joining pathway, which preferentially mediates repair of double-stranded breaks. Mutations in this gene cause different kinds of severe combined immunodeficiency disorders.
Specifications
| XLF | |
| Polyclonal | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| Q9H9Q4 | |
| NHEJ1 | |
| Synthetic peptide corresponding to amino acids near the carboxy terminus of human XLF. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| NHEJ1 | |
| 1700029B21Rik; cernunnos; FLJ12610; Nhej1; non-homologous end joining factor 1; non-homologous end-joining factor 1; nonhomologous end-joining factor 1; protein cernunnos; Xlf; XRCC4-like factor | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 79840 | |
| -20°C | |
| Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction