Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XP32 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306224F
Description
XP32 Polyclonal specifically detects XP32 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
XP32 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
C1orf68, chromosome 1 open reading frame 68, LEP7, skin-specific protein (xp32), skin-specific protein 32, XP32 | |
Produced in rabbits immunized with E. coli-derived Human XP32 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
FITC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
100129271 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction