Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XPA Antibody (2A4), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250600MFV450
Description
XPA Monoclonal antibody specifically detects XPA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| XPA | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| 2A4 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| DNA repair protein complementing XP-A cells, excision repair-controlling, Xeroderma pigmentosum group A-complementing protein, xeroderma pigmentosum, complementation group A, XPACXP1 | |
| His-tagged recombinant human XPA protein. | |
| 0.1 mL | |
| DNA Repair, Nucleotide Excision Repair | |
| 7507 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction