Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XPA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527852
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527852 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527852 Supplier Invitrogen™ Supplier No. PA527852
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T. Predicted reactivity: Mouse (88%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The XPA (xeroderma pigmentosumgroup A) protein specifically recognizes the UV-orchemically damaged DNA lesions, and triggers thenucleotide excision repair process. XPA binds to thereplication protein A (RPA) or the excision repaircross complementing 1 protein (ERCC 1). In the absence of nucleotide excision repair persisting (unrepaired) DNA lesions (adducts) may lead to the accumulation of gene mutations and ultimately to cancer. Xeroderma pigmentosum patients have a >2000 fold increased risk to develop skin cancer atsun-exposed areas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XPA
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene XPA
Gene Accession No. P23025
Gene Alias AI573865; DNA repair protein complementing XP-A cells; DNA repair protein complementing XP-A cells homolog; excision repair-controlling; mutant xeroderma pigmentosum complementation group A; xeroderma pigmentosum group A-complementing protein; Xeroderma pigmentosum group A-complementing protein homolog; xeroderma pigmentosum, complementation group A; XP1; XPA; XPA, DNA damage recognition and repair factor; Xpac
Gene Symbols XPA
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human XPA. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7507
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.