Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XPB Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10061059AF750
Description
XPB Polyclonal antibody specifically detects XPB in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
XPB | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Basic transcription factor 2 89 kDa subunit, BTF2, BTF2 p89, DNA excision repair protein ERCC-3, DNA repair protein complementing XP-B cells, EC 3.6.1, EC 3.6.4.12, excision repair cross-complementing rodent repair deficiency, complementationgroup 3 (xeroderma pigmentosum group B complementing), GTF2H, RAD25, TFIIH, TFIIH basal transcription factor complex 89 kDa subunit, TFIIH basal transcription factor complex helicase XPB subunit, TFIIH p89, Xeroderma pigmentosum group B-complementing protein, xeroderma pigmentosum, complementation group B, XPBC, XPBTFIIH 89 kDa subunit | |
The immunogen recognized by this antibody maps to a region between residue 575 and 625 of human excision repair cross-complementing rodent repair deficiency, complementation group 3 using the numbering given in entry NP_000113.1 (GeneID 2071). | |
0.1 mL | |
Cancer, DNA Repair, Nucleotide Excision Repair | |
2071 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction