Learn More
Invitrogen™ XPB Polyclonal Antibody

Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Specifications
Specifications
| Antigen | XPB |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.2 |
| Gene | Ercc3 |
| Gene Accession No. | P19447, P49135 |
| Gene Alias | Basic transcription factor 2 89 kDa subunit; BTF2; BTF2 p89; BTF2-p89; DNA excision repair protein ERCC-3; DNA repair helicase; DNA repair protein complementing XP-B cells; ERCC excision repair 3, TFIIH core complex helicase subunit; Ercc3; Ercc-3; excision repair 3; excision repair cross-complementation group 3; excision repair cross-complementing rodent repair deficiency, complementation group 3; General transcription and DNA repair factor IIH helicase subunit XPB; GTF2H; RAD25; TFIIH; TFIIH 89 kDa subunit; TFIIH basal transcription factor complex 89 kDa subunit; TFIIH basal transcription factor complex helicase XPB subunit; TFIIH p89; TFIIH subunit XPB; TTD2; Xeroderma pigmentosum group B-complementing protein; xeroderma pigmentosum, complementation group B; XPB; XPBC |
| Gene Symbols | Ercc3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.