Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ XPD Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528916
Description
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (96%), Rat (96%), Chimpanzee (99%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
XPD is involved with the nucleotide excision repair pathway that is a mechanism to repair damage to DNA. The protein encoded by this gene is involved in transcription-coupled nucleotide excision repair and is an integral member of the basal transcription factor BTF2/TFIIH complex. The gene product has ATP-dependent DNA helicase activity and belongs to the RAD3/XPD subfamily of helicases. Defects in this gene can result in three different disorders, the cancer-prone syndrome xeroderma pigmentosum complementation group D, trichothiodystrophy, and Cockayne syndrome. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
XPD | |
Polyclonal | |
Unconjugated | |
ERCC2 | |
AA407812; AU020867; AW240756; basic transcription factor 2 80 kDa subunit; BTF2 p80; COFS2; CXPD; DNA excision repair protein ERCC-2; DNA repair protein complementing XP-D cells; EM9; ERCC excision repair 2, TFIIH core complex helicase subunit; Ercc2; Ercc-2; excision repair cross-complementation group 2; excision repair cross-complementing rodent repair deficiency, complementation group 2; General transcription and DNA repair factor IIH helicase subunit XPD; MGC102762; MGC126218; MGC126219; TFIIH; TFIIH 80 kDa subunit; TFIIH basal transcription factor complex 80 kDa subunit; TFIIH basal transcription factor complex helicase subunit; TFIIH basal transcription factor complex helicase XPB subunit; TFIIH basal transcription factor complex helicase XPD subunit; TFIIH p80; TFIIH subunit XPD; TTD; TTD1; xeroderma pigmentosum complementary group D; xeroderma pigmentosum group D-complementing protein; XPD; XPDC | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2068 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P18074 | |
ERCC2 | |
Recombinant protein encompassing a sequence within the center region of human XPD. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction