Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XPE/DDB2 Antibody (24E5), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | XPE/DDB2 |
---|---|
Clone | 24E5 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
XPE/DDB2 Monoclonal antibody specifically detects XPE/DDB2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunohistochemistrySpecifications
XPE/DDB2 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
damage-specific DNA binding protein 2 (48kD), damage-specific DNA binding protein 2, 48kDa, Damage-specific DNA-binding protein 2, DDB p48 subunit, DDBb, DNA damage-binding protein 2, FLJ34321, UV-damaged DNA-binding protein 2, UV-DDB 2, UV-DDB2, xeroderma pigmentosum group E protein | |
A synthesized peptide derived from Human XPE/DDB2 [UniProt Q92466] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
24E5 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
DNA Repair, Nucleotide Excision Repair | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
1643 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title