Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XPE/DDB2 Rabbit anti-Human, Mouse, Clone: 4Y2G5, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | XPE/DDB2 |
---|---|
Clone | 4Y2G5 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
XPE/DDB2 Monoclonal antibody specifically detects XPE/DDB2 in Human, Mouse samples. It is validated for Western BlotSpecifications
XPE/DDB2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
DNA Repair, Nucleotide Excision Repair | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
1643 | |
IgG | |
Affinity purified |
4Y2G5 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
damage-specific DNA binding protein 2 (48kD), damage-specific DNA binding protein 2, 48kDa, Damage-specific DNA-binding protein 2, DDB p48 subunit, DDBb, DNA damage-binding protein 2, FLJ34321, UV-damaged DNA-binding protein 2, UV-DDB 2, UV-DDB2, xeroderma pigmentosum group E protein | |
Recombinant fusion protein containing a sequence corresponding to amino acids 220-427 of human XPE/DDB2 (Q92466). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title