Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Xpect™ RSV Test

Catalog No. r24601
Click to view available options
Quantity:
30 Tests
This item is not returnable. View return policy

Catalog No. R24601

Supplier: Thermo Scientific™ R24601

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rapidly detect Respiratory Syncytial Virus (RSV) from nasopharyngeal specimens in neonatal and pediatric patients and get results in 15 minutes.

Optimize your workflow by rapidly detecting Respiratory Syncytial Virus antigen (RSV) directly from nasopharyngeal specimens in neonatal and pediatric patients using Thermo Scientific™ Xpect™ RSV Test. Combine excellent clinical performance with a simple, 2-step procedure that offers accurate results within 15 minutes.

Use this rapid, lateral flow immunoassay for the direct qualitative detection of Respiratory Syncytial Virus (RSV). Reduce secondary testing, decrease unnecessary antibiotic use and guide appropriate antiviral therapy.
  • Improve patient management with accurate, easy-to-read results in just 15 minutes
  • Eliminate workflow bottlenecks with less than one minute hands-on time and a simple, walk-away procedure
  • Stock up with 18-month shelf life at room-temperature storage
  • Offers flexible specimen storage, collection, and transport options

Xpect RSV uses antibody-coated color particles to bind with target antigen in the sample. The color-antibody/antigen complex moves along the test membrane. Immobilised antibodies specific to target antigen capture the color-labelled antigen complex at the test region, causing a clearly visible line to appear. This line will not appear in the absence of specific antigen. Immobilised, non-specific antibodies in the control region capture the color-labelled antibodies, whether antigen is present or not, resulting in a clearly visible control line.

Trust the entire family of Xpect rapid tests to deliver unparalleled diagnostic performance while offering ease-of-use, convenience and time savings. Each test is an immunochromatographic assay with a simple procedure that provides rapid, cost-effective detection to aid in the diagnosis of infections.

Xpect Flu A&B
Xpect RSV
Xpect C. difficile Toxin A/B
Xpect Rotavirus
Xpect Giardia
Xpect Cryptosporidium
Xpect Giardia/Cryptosporidium
Xpect Legionella

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Description Xpect RSV Test
Detectable Analytes Respiratory Syncytial Virus (RSV) Antigen
Sample Type Nasopharyngeal
Type Reagent
Includes Test devices, extraction buffer (2 x 10mL), disposable extraction tubes, disposable pipettes
Quantity 30 Tests
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.