Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XPF Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | XPF |
---|---|
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
XPF Polyclonal specifically detects XPF in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
XPF | |
Polyclonal | |
Rabbit | |
Cancer, DNA Repair, Nucleotide Excision Repair | |
DNA excision repair protein ERCC-4, DNA repair protein complementing XP-F cells, EC 3.1, ERCC11, excision repair cross-complementing rodent repair deficiency, complementationgroup 4, xeroderma pigmentosum, complementation group F, XPFcomplementing defective, in Chinese hamster | |
ERCC4 | |
IgG | |
Affinity Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
2072 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SKPQPDAATALAITADSETLPESEKYNPGPQDFLLKMPGVNAKNCRSLMHHVKNIAELAALSQDELTSILGNAANAKQLYDFIHTSFAEVVSKGKGK | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title