Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XPF Antibody (OTI4E11), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274883APC
Description
XPF Monoclonal specifically detects XPF in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
| XPF | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| ERCC4 | |
| Full length recombinant protein of human XPF (NP_005227) produced in HEK293 cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG2a |
| Flow Cytometry | |
| OTI4E11 | |
| Flow Cytometry | |
| DNA excision repair protein ERCC-4, DNA repair protein complementing XP-F cells, EC 3.1, ERCC11, excision repair cross-complementing rodent repair deficiency, complementationgroup 4, xeroderma pigmentosum, complementation group F, XPFcomplementing defective, in Chinese hamster | |
| Mouse | |
| Affinity Purified | |
| Cancer, DNA Repair, Nucleotide Excision Repair | |
| 2072.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction