Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XPF Antibody (OTI4E11), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274883C
Description
XPF Monoclonal specifically detects XPF in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
XPF | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ERCC4 | |
Full length recombinant protein of human XPF (NP_005227) produced in HEK293 cell. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG2a |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI4E11 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
DNA excision repair protein ERCC-4, DNA repair protein complementing XP-F cells, EC 3.1, ERCC11, excision repair cross-complementing rodent repair deficiency, complementationgroup 4, xeroderma pigmentosum, complementation group F, XPFcomplementing defective, in Chinese hamster | |
Mouse | |
Affinity Purified | |
Cancer, DNA Repair, Nucleotide Excision Repair | |
2072.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction