Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XPF/ERCC4 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501571783
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1783 100 μL
50-157-1784 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-1783 Supplier Bethyl Laboratories, Inc Supplier No. IHC00263
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Monkey, Gorilla, Chimpanzee and Crab-eating macaque

The structure specific ERCC 1 / XPF endonuclease complex is implicated in the repair of two distinct types of lesions in DNA: NER for UV induced lesions and bulky chemical adducts; and recombination repair of the very genotoxic interstrand cross links. NER mechanism involves dual incisions on both sides of the damage catalyzed by two nucleases. In mammalian cells, XPG cleaves 3′ of the DNA lesion while the ERCC1 / XPF complex makes the 5′ incision. ERCC1 / XPF probably plays a role in processing partially processed recombination intermediates.

Specifications

Antigen XPF/ERCC4
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene ERCC4
Gene Accession No. Q92889
Gene Alias DNA excision repair protein ERCC-4, DNA repair endonuclease XPF, DNA repair protein complementing XP-F cells,excision-repair, complementing defective, in Chinese hamster, xeroderma pigmentosum group F-complementing protein, xeroderma pigmentosum, complementation group F
Gene Symbols ERCC4
Host Species Rabbit
Immunogen Between 855 and 905
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2072
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.