Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XPF Monoclonal Antibody (219), Invitrogen™

Catalog No. PIMA512054
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA512054 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA512054 Supplier Thermo Scientific Supplier No. MA512054

Mouse Monoclonal Antibody

MA5-12054 targets XPF in IHC (P) and WB applications and shows reactivity with Human samples. The MA5-12054 immunogen is recombinant human XPF protein.

XPF Monoclonal antibody specifically detects XPF in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XPF
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 219
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene ERCC4
Gene Accession No. Q92889
Gene Alias AI606920; DNA excision repair protein ERCC-4; DNA repair endonuclease subunit; DNA repair endonuclease XPF; DNA repair protein complementing XP-F cells; ERCC excision repair 4, endonuclease catalytic subunit; ERCC11; ERCC4; Ercc4 protein-like; excision repair cross-complementation group 4; excision repair cross-complementing rodent repair deficiency, complementation group 4; excision-repair, complementing defective, in Chinese hamster; FANCQ; RAD1; RGD1560340; Xeroderma pigmentosum group F-complementing protein; xeroderma pigmentosum, complementation group F; XFEPS; XPF
Gene Symbols ERCC4
Host Species Mouse
Immunogen Recombinant human XPF protein
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2072
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG2 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.