Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XPF Monoclonal Antibody (OTI2A1), TrueMAB™, OriGene
SDP

Catalog No. 501674240
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-4240 100 μL
50-254-0983 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-4240 Supplier OriGene Supplier No. TA503314
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The structure specific ERCC 1 / XPF endonuclease complex is implicated in the repair of two distinct types of lesions in DNA: NER for UV induced lesions and bulky chemical adducts; and recombination repair of the very genotoxic interstrand cross links. NER mechanism involves dual incisions on both sides of the damage catalyzed by two nucleases. In mammalian cells, XPG cleaves 3′ of the DNA lesion while the ERCC1 / XPF complex makes the 5′ incision. ERCC1 / XPF probably plays a role in processing partially processed recombination intermediates.

Specifications

Antigen XPF
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone OTI2A1
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ERCC4
Gene Accession No. Q92889, Q9QZD4
Gene Alias ERCC11, FANCQ, RAD1, XFEPS, XPF
Gene Symbols Ercc4
Host Species Mouse
Immunogen Full length human recombit protein of human ERCC4 produced in HEK293 cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2072, 304719, 479842, 50505
Target Species Canine, Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.