Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XPF Monoclonal Antibody (OTI2D4), TrueMAB™, OriGene
SDP

Catalog No. 502541140
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1140 30 μL
50-167-4035 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1140 Supplier OriGene Supplier No. TA503063S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

XPF Monoclonal antibody specifically detects XPF in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen XPF
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI2D4
Concentration 0.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Aldob
Gene Accession No. P05062
Gene Alias AI606920; DNA excision repair protein ERCC-4; DNA repair endonuclease subunit; DNA repair endonuclease XPF; DNA repair protein complementing XP-F cells; ERCC excision repair 4, endonuclease catalytic subunit; ERCC11; ERCC4; Ercc4 protein-like; excision repair cross-complementation group 4; excision repair cross-complementing rodent repair deficiency, complementation group 4; excision-repair, complementing defective, in Chinese hamster; FANCQ; RAD1; RGD1560340; Xeroderma pigmentosum group F-complementing protein; xeroderma pigmentosum, complementation group F; XFEPS; XPF
Host Species Mouse
Immunogen Full length human recombinant protein of human ALDOB produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 229
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.