Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XPF Monoclonal Antibody (OTI3H7), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503163S


Description
XPF Monoclonal antibody specifically detects XPF in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
XPF | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P53667 | |
Mouse | |
Affinity chromatography | |
RUO | |
3984 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI3H7 | |
Unconjugated | |
LIMK1 | |
AI606920; DNA excision repair protein ERCC-4; DNA repair endonuclease subunit; DNA repair endonuclease XPF; DNA repair protein complementing XP-F cells; ERCC excision repair 4, endonuclease catalytic subunit; ERCC11; ERCC4; Ercc4 protein-like; excision repair cross-complementation group 4; excision repair cross-complementing rodent repair deficiency, complementation group 4; excision-repair, complementing defective, in Chinese hamster; FANCQ; RAD1; RGD1560340; Xeroderma pigmentosum group F-complementing protein; xeroderma pigmentosum, complementation group F; XFEPS; XPF | |
Full length human recombinant protein of human LIMK1 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction