Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XPF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117118
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117118 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117118 Supplier Invitrogen™ Supplier No. PA5117118
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: U-251MG, HT-1080 Immunogen sequence: GYRREISSSP ESCPEEIKHE EFDVNLSSDA AFGILKEPLT IIHPLLGCSD PYALTRVLHE VEPRYVVLYD AELTFVRQLE IYRASRPGKP LRVYFLIYGG STEEQRYLTA LRKEKEAFEK LIREKASMVV PEEREGRDET NLDLVRGTAS ADVSTDTRKA GGQEQNGTQQ SIVVDMREFR SELPSLIHRR GIDIEPVTLE VGDYILTPEM CVERKSISDL IGSLNNGRLY SQCISMSRYY KRPVLLIEFD PSKPFSLTSR GALFQEISSN.

The structure specific ERCC 1 / XPF endonuclease complex is implicated in the repair of two distinct types of lesions in DNA: NER for UV induced lesions and bulky chemical adducts; and recombination repair of the very genotoxic interstrand cross links. NER mechanism involves dual incisions on both sides of the damage catalyzed by two nucleases. In mammalian cells, XPG cleaves 3' of the DNA lesion while the ERCC1 / XPF complex makes the 5' incision. ERCC1 / XPF probably plays a role in processing partially processed recombination intermediates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XPF
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ERCC4
Gene Accession No. Q92889, Q9QZD4
Gene Alias AI606920; DNA excision repair protein ERCC-4; DNA repair endonuclease subunit; DNA repair endonuclease XPF; DNA repair protein complementing XP-F cells; ERCC excision repair 4, endonuclease catalytic subunit; ERCC11; ERCC4; Ercc4 protein-like; excision repair cross-complementation group 4; excision repair cross-complementing rodent repair deficiency, complementation group 4; excision-repair, complementing defective, in Chinese hamster; FANCQ; RAD1; RGD1560340; Xeroderma pigmentosum group F-complementing protein; xeroderma pigmentosum, complementation group F; XFEPS; XPF
Gene Symbols ERCC4
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2072, 304719, 50505
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.