Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XPG Antibody (8H7/XPG), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250366C
Description
XPG Monoclonal specifically detects XPG in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry.Specifications
XPG | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ERCC5 | |
Recombinant human XPG protein produced in baculovirus | |
0.1 mL | |
Cancer, DNA Repair, Nucleotide Excision Repair | |
2073 | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
8H7/XPG | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
COFS3, DNA excision repair protein ERCC-5, DNA repair protein complementing XP-G cells, EC 3.1, ERCM2, excision repair cross-complementing rodent repair deficiency, complementationgroup 5, excision repair protein, xeroderma pigmentosum complementation group G protein, Xeroderma pigmentosum group G-complementing protein, xeroderma pigmentosum, complementation group G, XPGC, XPG-complementing protein, XPGUVDR | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction