Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XPG Monoclonal Antibody (8H7)

Catalog No. PIMA126186
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA126186 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA126186 Supplier Invitrogen™ Supplier No. MA126186

Mouse Monoclonal Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Xeroderma pigmentosum type G (XPG) is a human genetic disease exhibiting extreme sensitivity to sunlight. The XPG protein, a member of the flap endonuclease 1 (FEN-1) structure-specific DNA repair endonuclease family, is an enzyme essential for DNA repair of the major kinds of solar ultraviolet (UV)-induced DNA damages. Human XPG nuclease makes the 3' incision during nucleotide excision repair of DNA. The enzyme cleaves model DNA bubble structures specifically near the junction of unpaired DNA with a duplex region. A 29-amino acid region of human XPG (residues 981-1009) contains the PCNA binding activity. A conserved arginine in XPG (Arg992) is crucial for its PCNA binding activity. Replication Protein A (RPA) binds specifically and directly to two excision repair proteins, the xeroderma pigmentosum damage-recognition protein XPA and the endonuclease XPG.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XPG
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 8H7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ERCC5
Gene Accession No. P28715
Gene Alias COFS 3; COFS3; DNA excision repair protein ERCC-5; DNA repair protein complementing XP-G cells; DNA repair protein complementing XP-G cells homolog; ERCC 5; ERCC excision repair 5, endonuclease; ERCC5; ERCC5-201; ERCM 2; ERCM2; excision repair cross-complementation group 5; excision repair cross-complementing rodent repair deficiency, complementation group 5; OTTHUMP00000064902; RP11-484I6.5; UVDR; Xe; xeroderma pigmentosum; Xeroderma pigmentosum group G-complementing protein; xeroderma pigmentosum group G-complementing protein homolog; xeroderma pigmentosum, complementation group G; XPG; XPGC; XPG-complementing protein
Gene Symbols ERCC5
Host Species Mouse
Immunogen Recombinant human XPG (Xeroderma Pigmentosum type G) protein produced in Baculovirus.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2073
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.