Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Xplorer™ plus Electronic Pipette, Multi-Channel PROMO

Catalog No. 14286558PM Shop All Eppendorf North America Products
Change view
Click to view available options
Eppendorf™ Xplorer™ plus Electronic Pipette
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
14-286-558PM Eppendorf™ Xplorer™ plus Electronic Pipette
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-286-558PM Supplier Eppendorf™ Supplier No. 2231001121

Multi-channel electronic pipette with intuitive operating system and design for simplicity, precision and reproducibility.

Facilitates performance of additional, intelligent applications including: aspirating, diluting, sequential dispensing and reverse pipetting. Also, helps eliminate delays due to complicated programming and/or inflexible processes. Provides precisely adjustable parameters, maximum reproducible results, fatigue-free work and consistent, full control over the pipetting processes.

  • Multifunctional rocker: Up is up and down is down™ principle
  • Easy selection of all functions with the practical selection dial
  • Operate in the language of your choice
  • Clearly arranged color display; no sub-menus; help function
  • Innovative rocker: When the tip is ejected, the piston automatically returns to zero position
  • ON/OFF switch: Switches the pipetter off during longer periods of inactivity
  • Additional applications include aspirating, diluting, sequential dispensing and reverse pipetting
  • Programming: One-time programming of sequences for easier handling of complex processes
  • Individual fixed volume setting: Desired volume can be saved
  • Password protection: Protects programming and adjustment
  • Individual speed setting: Sets parameters to your individual working speed
  • Pipetter maintenance reminder function, based on time or frequency of use as desired
Reliably reproducible results
  • Tips always rest in the same position
  • Volume and speed precisely and reproducibly adjustable
  • Adjustable to your work environment and requirements
PhysioCare Concept™
  • Reduced weight
  • Improved ergonomics
  • Perfect balance
  • Optimally angled pipetter head
  • Improved view and high resolution display
  • Spring-loaded tip cone provides maximum tightness
Applications
  • Pipetting liquids (manual and automatic)
  • Dispensing (timed and manual)
  • Pipetting and mixing
TRUSTED_SUSTAINABILITY

Specifications

Battery Type Lithium-ion
Compatible Tips 200 μL Tip
Color Yellow
Disposable Reusable
For Use With Pipette Tip (200 μL)
Number of Channels 8
Product Line Xplorer Plus
Programmability Yes
Type Electronic Pipette
Volume (Metric) 5 to 100 μL
Display LCD
Finger Style Ergonomic
For Use With (Equipment) 200 μL Tip
Includes 10 Pipetting Modes, Pipetting History, Password Protection, a Service Indicator and the Ability to Program Multiple Modes in a Sequence for Custom Applications
Increments 0.1 μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.