Learn More
XPNPEP1 Mouse anti-Human, Clone: OTI1E7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501384

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes the cytosolic form of a metalloaminopeptidase that catalyzes the cleavage of the N-terminal amino acid adjacent to a proline residue. The gene product may play a role in degradation and maturation of tachykinins, neuropeptides, and peptide hormones. Alternative splicing results in multiple transcript variants.Specifications
XPNPEP1 | |
Monoclonal | |
Unconjugated | |
XPNPEP1 | |
APP1, SAMP, XPNPEP, XPNPEPL, XPNPEPL1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
7511 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1E7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9NQW7 | |
XPNPEP1 | |
Full length human recombit protein of human XPNPEP1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.