Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XPNPEP1 Recombinant Mouse Monoclonal Antibody (4D12)
GREENER_CHOICE

Catalog No. FEMA550245
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550245 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550245 Supplier Invitrogen™ Supplier No. MA550245
Add to Cart
Add to Cart

Mouse Recombinant Monoclonal Antibody

XPNPEP1 Recombinant Monoclonal Antibody for IHC (P), Flow, ELISA

This gene encodes the cytosolic form of a metalloaminopeptidase that catalyzes the cleavage of the N-terminal amino acid adjacent to a proline residue. The gene product may play a role in degradation and maturation of tachykinins, neuropeptides, and peptide hormones. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XPNPEP1
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone 4D12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Xpnpep1
Gene Accession No. Q9NQW7
Gene Alias Aminoacylproline aminopeptidase; aminopeptidase P, cytosolic; App; APP1; cytoplasmic aminopeptidase P; cytosolic aminopeptidase P; D230045I08Rik; OTTHUMP00000020457; OTTHUMP00000058856; RGD:621274}; sAmp; soluble aminopeptidase P; Xaa-Pro aminopeptidase 1; XPNPEP; XPNPEP1; xpnpep1 {ECO:0000312; XPNPEPL; XPNPEPL1; X-Pro aminopeptidase 1; X-Pro aminopeptidase 1, soluble; X-prolyl aminopeptidase (aminopeptidase P) 1, soluble; X-prolyl aminopeptidase 1; X-prolyl aminopeptidase 1, soluble
Gene Symbols Xpnpep1
Host Species Mouse
Immunogen Recombinant Human XPNPEP1 protein.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7511
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.