Learn More
Invitrogen™ XPNPEP1 Recombinant Rabbit Monoclonal Antibody (SAIC-07E-1)

Description
Specificity: Recognises human XPNPEP1.
Specifications
Specifications
| Antigen | XPNPEP1 |
| Applications | ELISA, Flow Cytometry, Western Blot, Immunocytochemistry, Mass Spectrometry |
| Classification | Recombinant Monoclonal |
| Clone | SAIC-07E-1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Gene | Xpnpep1 |
| Gene Accession No. | Q9NQW7 |
| Gene Alias | Aminoacylproline aminopeptidase; aminopeptidase P, cytosolic; App; APP1; cytoplasmic aminopeptidase P; cytosolic aminopeptidase P; D230045I08Rik; OTTHUMP00000020457; OTTHUMP00000058856; RGD:621274}; sAmp; soluble aminopeptidase P; Xaa-Pro aminopeptidase 1; XPNPEP; XPNPEP1; xpnpep1 {ECO:0000312; XPNPEPL; XPNPEPL1; X-Pro aminopeptidase 1; X-Pro aminopeptidase 1, soluble; X-prolyl aminopeptidase (aminopeptidase P) 1, soluble; X-prolyl aminopeptidase 1; X-prolyl aminopeptidase 1, soluble |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.