Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XPNPEP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587752
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587752 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587752 Supplier Invitrogen™ Supplier No. PA587752
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HTKPVDLGGQ DVRNCSTNPP YLPVTVVNTT MSLTALRQQM QTQNLSAYII PGTDAHMNEY IGQHDERRAW ITGFTGSAGT AVVTMKKAAV WTDSRYWTQA ERQMDCNWEL HKEVGTTPIV TWLLTEIPAG GRVGFDPFLL SIDTWESYDL ALQGSNRQLV SITTNLVDLV WGSERPPVPN QPIYALQEAF TGSTWQEKVS GVRSQMQKHQ KVPTAVLLS; Positive Samples: 293T, A-549, HepG2, HT-29, SW480, Mouse kidney, Mouse heart, Mouse small intestine, Rat kidney; Cellular Location: Cell membrane, GPI-anchor, Lipid-anchor.

Aminopeptidase P is a hydrolase specific for N-terminal imido bonds, which are common to several collagen degradation products, neuropeptides, vasoactive peptides, and cytokines. Structurally, the enzyme is a member of the 'pita bread fold' family and occurs in mammalian tissues in both soluble and GPI-anchored membrane-bound forms. A membrane-bound and soluble form of this enzyme have been identified as products of two separate genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XPNPEP2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.06 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene XPNPEP2
Gene Accession No. B1AVD1, O43895, Q99MA2
Gene Alias 9030008G12Rik; AEACEI; Aminoacylproline aminopeptidase; APP2; LOW QUALITY PROTEIN: xaa-Pro aminopeptidase 2; mAmP; mAPP; membrane-bound aminopeptidase P; membrane-bound AmP; membrane-bound APP; RP4-753P9.2; xaa-Pro aminopeptidase 2; Xpnpep2; X-Pro aminopeptidase 2; X-prolyl aminopeptidase (aminopeptidase P) 2, membrane-bound; X-prolyl aminopeptidase 2; X-prolyl aminopeptidase 2, membrane-bound
Gene Symbols XPNPEP2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 22-240 of human XPNPEP2 (NP_0033904).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117522, 170745, 7512
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.