Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XPO4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565820
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565820 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565820 Supplier Invitrogen™ Supplier No. PA565820
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DIHWLILVTGY LLADDTQGET PLIPPEIMEY SIKHSSEVDI NTTLQILGSP GEKASSIPGY NRTDSVIRLL SAILRVSEVE SRAIRADLTH LLSPQMG Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.

Nuclear transport receptors of karyopherin/importin-beta superfamily mediate various transport pathways between nucleus and cytoplasm. Exportin-4 is a novel mammalian member of this family. It is a RanGTP-binding protein consisting of N-terminal RanGTP-binding motif. It functions as a nuclear export receptor for eIF-5A and also might function as an export adapter in RNA export. It is also involved in nuclear export of Smad3 and regulation of Smad signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XPO4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene XPO4
Gene Accession No. Q9C0E2
Gene Alias B430309A01Rik; exp4; exportin 4; exportin-4; FLJ13046; hypothetical protein; KIAA1721; mKIAA1721; RCJMB04_1f17; Xpo4
Gene Symbols XPO4
Host Species Rabbit
Immunogen Recombinant Human XPO4. Recombinant protein control fragment (Product #RP-106493).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64328
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.