Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XPO5 Recombinant Rabbit Monoclonal Antibody (3Q8E6)
GREENER_CHOICE

Catalog No. PIMA535406
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535406 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535406 Supplier Thermo Scientific Supplier No. MA535406
Add to Cart
Add to Cart

XPO5 Recombinant Rabbit Monoclonal Antibody (ARC0838)

XPO5 Recombinant Monoclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen XPO5
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 3Q8E6
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene XPO5
Gene Accession No. Q924C1, Q9HAV4
Gene Alias 2410004H11Rik; 2700038C24Rik; AI648907; AW549301; exp5; exportin 5; exportin-5; FLJ14239; FLJ32057; FLJ45606; KIAA1291; mKIAA1291; Ran-binding protein 21; RanBp21; Xpo5
Gene Symbols XPO5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human Exportin 5 (XPO5) (Q9HAV4).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 363194, 57510, 72322
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.