Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XPO5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554941
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554941 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554941 Supplier Invitrogen™ Supplier No. PA554941
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASLVHLAFQI YEALRPRYLE IRAVMEQIPE IQKDSLDQFD CKLLNPSLQK VADKRRKDQF KRLIAGCIGK PLGEQFRKEV H Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

Exportin-5 belongs to a large family of karyopherins that mediate the transport of proteins and other cargo between the nuclear and cytoplasmic compartments. Exportin-5 belongs to a large family of karyopherins that mediate the transport of proteins and other cargo between the nuclear and cytoplasmic compartments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XPO5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene XPO5
Gene Accession No. Q924C1, Q9HAV4
Gene Alias 2410004H11Rik; 2700038C24Rik; AI648907; AW549301; exp5; exportin 5; exportin-5; FLJ14239; FLJ32057; FLJ45606; KIAA1291; mKIAA1291; Ran-binding protein 21; RanBp21; Xpo5
Gene Symbols XPO5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human XPO5. Recombinant protein control fragment (Product #RP-91844).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 363194, 57510, 72322
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.