Learn More
Invitrogen™ XPO5 Polyclonal Antibody

Description
Immunogen sequence: ILGPLFTYLH MRLSQKWQVI NQRSLLCGED EAADENPESQ EMLEEQLVRM LTREVMDLIT VCCVSKKGAD HSSAPPADGD DEEMMATEVT PSAMAELTDL GKCLMKHEDV CTALLITAFN SLAWKDTLSC Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 86%.
Specifications
Specifications
| Antigen | XPO5 |
| Applications | Western Blot, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | XPO5 |
| Gene Accession No. | Q924C1, Q9HAV4 |
| Gene Alias | 2410004H11Rik; 2700038C24Rik; AI648907; AW549301; exp5; exportin 5; exportin-5; FLJ14239; FLJ32057; FLJ45606; KIAA1291; mKIAA1291; Ran-binding protein 21; RanBp21; Xpo5 |
| Gene Symbols | XPO5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.