Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ XPO5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556293
Description
Immunogen sequence: MSFCVYSILG VVKRTCWPTD LEEAKAGGFV VGYTSSGNPI FRNPCTEQIL KLLDNLLALI RTHNTLYAPE MLAKMAEPFT KALDMLDAEK SAILGLPQPL LELNDSPVFK TVLERMQRFF STLYENC Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.
Exportin-5 belongs to a large family of karyopherins that mediate the transport of proteins and other cargo between the nuclear and cytoplasmic compartments. Exportin-5 belongs to a large family of karyopherins that mediate the transport of proteins and other cargo between the nuclear and cytoplasmic compartments.
Specifications
XPO5 | |
Polyclonal | |
Unconjugated | |
XPO5 | |
2410004H11Rik; 2700038C24Rik; AI648907; AW549301; exp5; exportin 5; exportin-5; FLJ14239; FLJ32057; FLJ45606; KIAA1291; mKIAA1291; Ran-binding protein 21; RanBp21; Xpo5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57510, 72322 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q924C1, Q9HAV4 | |
XPO5 | |
Recombinant protein corresponding to Human XPO5. Recombinant protein control fragment (Product #RP-91842). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction