Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XPO6 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10059853V
Description
XPO6 Polyclonal antibody specifically detects XPO6 in Human samples. It is validated for Western BlotSpecifications
XPO6 | |
Polyclonal | |
Western Blot | |
Exp6, exportin 6, exportin-6, FLJ22519, KIAA0370EXP6, RAN binding protein 20, Ran-binding protein 20, RANBP20 | |
The immunogen recognized by this antibody maps to a region between residue 1075 and 1125 of human exportin 6 using the numbering given in entry NP_055986.1 (GeneID 23214). | |
0.1 mL | |
Signal Transduction | |
23214 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction