Learn More
XPO6/Exportin 6 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301205AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and X. laevis
Exportins, such as XPO6, recruit cargo in the nucleoplasm in the presence of RAN (MIM 601179)-GTP and form ternary export complexes. These complexes are transported through nuclear pore complexes to the cytoplasm, where GTP is hydrolyzed and the export complex is disassembled.Specifications
| XPO6/Exportin 6 | |
| Polyclonal | |
| Unconjugated | |
| XPO6 | |
| EXP6, RANBP20, RAN binding protein 20, exportin-6, ran-binding protein 20 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23214 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q96QU8 | |
| XPO6 | |
| Between 1075 and 1125 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.