Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ XPO6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531813
Description
Recommended positive controls: IMR32, U87-MG. Predicted reactivity: Mouse (97%), Rat (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a member of the importin-beta family. Members of this family are regulated by the GTPase Ran to mediate transport of cargo across the nuclear envelope. This protein has been shown to mediate nuclear export of profilin-actin complexes. A pseudogene of this gene is located on the long arm of chromosome 14. Alternative splicing results in multiple transcript variants that encode different protein isoforms.
Specifications
| XPO6 | |
| Polyclonal | |
| Unconjugated | |
| Xpo6 | |
| 2610005L19Rik; AI854665; AL022631; C230091E20Rik; Exp6; exportin 6; exportin-6; KIAA0370; mKIAA0370; R75304; RAN binding protein 20; ran-binding protein 20; Ranbp20; Xpo6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23214 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q96QU8 | |
| Xpo6 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human XPO6. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction