Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XPO6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558311
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558311 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558311 Supplier Invitrogen™ Supplier No. PA558311
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RPSPDVKAEL FELLFRTLHH NWRYFFKSTV LASVQRGIAE EQMENEPQFS AIMQAFGQSF LQPDIHLFKQ NLFYLETLNT KQKLYHKK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

The protein encoded by this gene is a member of the importin-beta family. Members of this family are regulated by the GTPase Ran to mediate transport of cargo across the nuclear envelope. This protein has been shown to mediate nuclear export of profilin-actin complexes. A pseudogene of this gene is located on the long arm of chromosome 14. Alternative splicing results in multiple transcript variants that encode different protein isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XPO6
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Xpo6
Gene Accession No. Q96QU8
Gene Alias 2610005L19Rik; AI854665; AL022631; C230091E20Rik; Exp6; exportin 6; exportin-6; KIAA0370; mKIAA0370; R75304; RAN binding protein 20; ran-binding protein 20; Ranbp20; Xpo6
Gene Symbols Xpo6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human XPO6. Recombinant protein control fragment (Product #RP-95685).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23214
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.