Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XPOT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588317
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588317 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588317 Supplier Invitrogen™ Supplier No. PA588317
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDEQALLGLN PNADSDFRQR ALAYFEQLKI SPDAWQVCAE ALAQRTYSDD HVKFFCFQVL EHQVKYKYSE LTTVQQQLIR ETLISWLQAQ MLNPQPEKTF IRNKAAQVFA LLFVTEYLTK WPKFFFDILS VVDLNPRGVD LYLRILMAID SELVDRDVVH TSEEARRNTL IKDTMREQCI PNLVESWYQI LQNYQFTNSE; Positive Samples: B-cell, Mouse brain, Mouse kidney, Mouse liver, Rat kidney; Cellular Location: Cytoplasm, Nucleus.

XPOT is a protein belonging to the RAN-GTPase exportin family that mediates export of tRNA from the nucleus to the cytoplasm. Translocation of tRNA to the cytoplasm occurs once exportin has bound both tRNA and GTP-bound RAN.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XPOT
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene XPOT
Gene Accession No. O43592, Q9CRT8
Gene Alias 1110004L07Rik; 3110065H13Rik; AI452076; C79645; exportin for tRNA; exportin(tRNA); exportin, tRNA; exportin, tRNA (nuclear export receptor for tRNAs); exportin-T; tRNA exportin; XPO3; XPOT
Gene Symbols XPOT
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human XPOT (NP_0091662).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11260, 314879, 73192
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.