Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XPV/DNA polymerase eta Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB10060424
Description
XPV/DNA polymerase eta Polyclonal specifically detects XPV/DNA polymerase eta in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
XPV/DNA polymerase eta | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
DNA polymerase eta, EC 2.7.7.7, polymerase (DNA directed), eta, RAD30, RAD30 homolog A, RAD30AFLJ16395, Xeroderma pigmentosum variant type protein, XP-V, XPVFLJ21978 | |
Rabbit | |
78 kDa | |
0.1 mL | |
DNA Polymerases, DNA Repair | |
5429 | |
Human | |
IgG |
Western Blot, Immunoprecipitation | |
0.2 mg/mL | |
Western Blot 1:2000 -1:10000, Immunoprecipitation 2-10 μg/mg lysate | |
Q9UQ35 | |
POLH | |
The immunogen recognized by this antibody maps to a region between residue 690 and 713 of human polymerase (DNA directed), eta using the numbering given in entry NP_006493.1 (GeneID 5429). | |
Affinity Purified | |
RUO | |
Primary | |
Based on 100% sequence identity, this antibody is predicted to react with Panda, Orangutan, Rhesus Monkey, Chimpanzee, White-tufted-ear marmoset, Crab-eating macaque, African elephant, Chinese hamster, Naked mole rat and Northern white-cheeked gibbon. | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction