Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ XRCC1 Monoclonal Antibody (7B4C12)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA548629
Description
XRCC1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow, ELISA
XRCC1 (X-ray cross complementing factor-1) is involved in DNA base excision repair. XRCC1 is a 70 kDa protein that has been shown to be physically associated with other DNA repair enzymes, including poly (ADP-ribose) polymerase (PARP), DNA Ligase III and DNA polymerase beta. XRCC1 contains a BRCT domain (for BRCA1 carboxyl terminus) which was originally found in the tumor suppressor protein BRCA1. Recently, the BRCT domain has been shown to mediate the protein-protein interaction of XRCC1 and DNA ligase III-alpha.
Specifications
XRCC1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P18887 | |
XRCC1 | |
Purified recombinant fragment of human XRCC1 (AA: 1-150) expressed in E. Coli. | |
100 μg | |
Primary | |
Human, Monkey | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
7B4C12 | |
Unconjugated | |
XRCC1 | |
DNA repair protein XRCC1; RCC; X-ray repair complementing defective repair in Chinese hamster cells 1; X-ray repair cross complementing 1; x-ray repair cross-complementing group 1 protein; X-ray repair cross-complementing protein 1; XRCC1; Xrcc-1 | |
Mouse | |
Protein G | |
RUO | |
7515 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction