Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XRCC1 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501991299
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1299 25 μL
50-199-1298 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-1299 Supplier Rockland Immunochemicals Supplier No. 200901MG5S
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This purified antibody is directed against human XRCC1. This product is an IgY fraction antibody purified from monospecific chicken egg yolks by a multi-step process which includes selective precipitation and salt fractionation followed by extensive dialysis against the buffer stated above.

XRCC1 (X-ray cross complementing factor-1) is involved in DNA base excision repair. XRCC1 is a 70 kDa protein that has been shown to be physically associated with other DNA repair enzymes, including poly (ADP-ribose) polymerase (PARP), DNA Ligase III and DNA polymerase beta. XRCC1 contains a BRCT domain (for BRCA1 carboxyl terminus) which was originally found in the tumor suppressor protein BRCA1. Recently, the BRCT domain has been shown to mediate the protein-protein interaction of XRCC1 and DNA ligase III-alpha.

Specifications

Antigen XRCC1
Applications ELISA, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation 0.002M sodium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7
Gene XRCC1
Gene Accession No. P18887, Q60596
Gene Alias DNA repair protein XRCC1; RCC; X-ray repair complementing defective repair in Chinese hamster cells 1; X-ray repair cross complementing 1; x-ray repair cross-complementing group 1 protein; X-ray repair cross-complementing protein 1; Xrcc1; Xrcc-1
Gene Symbols XRCC1
Host Species Chicken
Immunogen Anti-XRCC1 antibody was prepared from eggs of chickens laid after repeated immunizations with a synthetic peptide corresponding to a N-Terminal portion of human XRCC1 conjugated to Keyhole Limpet Hemocyanin (KLH).
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22594, 7515
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.