Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XRCC1 Rabbit anti-Human, Mouse, Rat, Clone: 2X1U4, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | XRCC1 |
---|---|
Clone | 2X1U4 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
XRCC1 Monoclonal antibody specifically detects XRCC1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
XRCC1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Base Excision Repair, Cancer, DNA Repair, Homologous Recombination, Tumor Suppressors | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
7515 | |
IgG | |
Affinity purified |
2X1U4 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
DNA repair protein XRCC1, RCC, X-ray repair complementing defective repair in Chinese hamster cells 1, X-ray repair cross-complementing protein 1, X-ray-repair, complementing defective, repair in Chinese hamster | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human XRCC1 (P18887). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title