Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XRCC3 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100165PCP
Description
XRCC3 Polyclonal antibody specifically detects XRCC3 in Human, Drosophila, Hamster samples. It is validated for Western Blot, Immunoprecipitation, ICC/IF (Negative)Specifications
XRCC3 | |
Polyclonal | |
Western Blot, Immunoprecipitation, ICC/IF (Negative) | |
CMM6, DNA repair protein XRCC3, X-ray repair complementing defective repair in Chinese hamster cells 3, X-ray repair cross-complementing protein 3 | |
A synthetic partial peptide of human XRCC3. [UniProt# O43542] | |
0.1 mL | |
Apoptosis, Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents, Homologous Recombination, Tumor Suppressors | |
7517 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
PerCP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Drosophila, Hamster | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction